Merck
  • A potential role for protein palmitoylation and zDHHC16 in DNA damage response.
A potential role for protein palmitoylation and zDHHC16 in DNA damage response.
BMC molecular biology (2016-05-11)
Na Cao, Jia-Kai Li, Yu-Qing Rao, Huijuan Liu, Ji Wu, Baojie Li, Peiquan Zhao, Li Zeng, Jing Li
ABSTRACT
Cells respond to DNA damage by activating the phosphatidylinositol-3 kinase-related kinases, p53 and other pathways to promote cell cycle arrest, apoptosis, and/or DNA repair. Here we report that protein palmitoylation, a modification carried out by protein acyltransferases with zinc-finger and Asp-His-His-Cys domains (zDHHC), is required for proper DNA damage responses. Inhibition of protein palmitoylation compromised DNA damage-induced activation of Atm, induction and activation of p53, cell cycle arrest at G2/M phase, and DNA damage foci assembly/disassembly in primary mouse embryonic fibroblasts. Furthermore, knockout of zDHHC16, a palmitoyltransferase gene identified as an interacting protein for c-Abl, a non-receptor tyrosine kinase involved in DNA damage response, reproduced most of the defects in DNA damage responses produced by the inhibition of protein palmitoylation. Our results revealed critical roles for protein palmitoylation and palmitoyltransferase zDHHC16 in early stages of DNA damage responses and in the regulation of Atm activation.
Sigma-Aldrich
Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12, clone 10H11.E12, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse