- Human articular chondrocytes express 15-lipoxygenase-1 and -2: potential role in osteoarthritis.
Human articular chondrocytes express 15-lipoxygenase-1 and -2: potential role in osteoarthritis.
15-Lipoxygenases and their metabolites have been shown to exhibit anti-inflammatory and immunomodulatory properties, but little is known regarding their expression and function in chondrocytes. The objective of this study was to evaluate the expression of 15-lipoxygenase-1 and -2 in human articular chondrocytes, and to investigate the effects of their metabolites 13(S)-hydroxy octadecadienoic and 15(S)-hydroxyeicosatetraenoic acids on IL-1beta-induced matrix metalloproteinase (MMP)-1 and MMP-13 expression. The expression levels of 15-lipoxygenase-1 and -2 were analyzed by reverse transcription PCR and Western blotting in chondrocytes, and by immunohistochemistry in cartilage. Chondrocytes or cartilage explants were stimulated with IL-1beta in the absence or presence of 13(S)-hydroxy octadecadienoic and 15(S)-hydroxyeicosatetraenoic acids, and the levels of MMP-1 and MMP-13 protein production and type II collagen cleavage were evaluated using immunoassays. The role of peroxisome proliferator-activated receptor (PPAR)gamma was evaluated using transient transfection experiments and the PPARgamma antagonist GW9662. Articular chondrocytes express 15-lipoxygenase-1 and -2 at the mRNA and protein levels. 13(S)-hydroxy octadecadienoic and 15(S)-hydroxyeicosatetraenoic acids dose dependently decreased IL-1beta-induced MMP-1 and MMP-13 protein and mRNA expression as well as type II collagen cleavage. The effect on MMP-1 and MMP-13 expression does not require de novo protein synthesis. 13(S)-hydroxy octadecadienoic and 15(S)-hydroxyeicosatetraenoic acids activated endogenous PPARgamma, and GW9662 prevented their suppressive effect on MMP-1 and MMP-13 production, suggesting the involvement of PPARgamma in these effects. This study is the first to demonstrate the expression of 15-lipoxygenase-1 and -2 in articular chondrocytes. Their respective metabolites, namely 13(S)-hydroxy octadecadienoic and 15(S)-hydroxyeicosatetraenoic acids, suppressed IL-1beta-induced MMP-1 and MMP-13 expression in a PPARgamma-dependent pathway. These data suggest that 15-lipoxygenases may have chondroprotective properties by reducing MMP-1 and MMP-13 expression.