Merck
  • Synthesis and Characterization of Core-shell ZrO2/PAAEM/PS Nanoparticles.
Synthesis and Characterization of Core-shell ZrO2/PAAEM/PS Nanoparticles.
Nanoscale research letters (2008-01-01)
J Wang, Tj Shi, Xc Jiang
ABSTRACT
This work demonstrates the synthesis of core-shell ZrO2/PAAEM/PS nanoparticles through a combination of sol-gel method and emulsifier-free emulsion polymerizaiton. By this method, the modified nanometer ZrO2cores were prepared by chemical modification at a molecular level of zirconium propoxide with monomer of acetoacetoxyethylmethacrylate (AAEM), and then copolymerized with vinyl monomer to form uniform-size hybrid nanoparticles with diameter of around 250 nm. The morphology, composition, and thermal stability of the core-shell particles were characterized by various techniques including transmission electron microscopy (TEM), X-ray diffractometer (XRD), Fourier transform infrared spectroscopy (FTIR), X-ray photoelectron spectroscopy (XPS), and thermal-gravimetry analyzer (TGA). The results indicate that the inorganic-organic nanocomposites exhibit good thermal stability with the maximum decomposition temperature of ~447 °C. This approach would be useful for the synthesis of other inorganic-organic nanocomposites with desired functionalities.
Sigma-Aldrich
2-(Methacryloyloxy)ethyl acetoacetate, 95%, contains BHT as stabilizer