Merck
  • Petatewalide B, a novel compound from Petasites japonicus with anti-allergic activity.
Petatewalide B, a novel compound from Petasites japonicus with anti-allergic activity.
Journal of ethnopharmacology (2015-12-18)
Young-Whan Choi, Kyoung-Pil Lee, Jung-Min Kim, Saeromi Kang, Soo-Jin Park, Jung-Min Lee, Hyung Ryong Moon, Jee H Jung, Young-Geun Lee, Dong-Soon Im
ABSTRACT
The giant butterbur Petasites japonicus is used to treat asthma and allergic diseases in traditional Korean, Japanese, and Chinese medicine. To elucidate the anti-allergic effect of Petasites genus, we studied effects of several compounds from Petasites japonicus leaves and found a novel bakkenolide-type sesquiterpine. In the present study, anti-allergic and anti-inflammatory effects of the new compound was examined using in vivo and in vitro experiments. The novel compound was isolated from Petasites japonicus leaves and named petatewalide B. Antigen-induced degranulation and Ca(2+) mobilization were measured in RBL-2H3 mast cells by measuring β-hexosaminidase activity and fluorescence change of Ca(2+) probe, fura-2. Induction of inducible nitric oxide synthase and cyclooxygenase 2 was measured by Western blotting in peritoneal macrophages. In addition, ovalbumin-induced asthma model was used for in vivo efficacy test of petatewalide B. Membrane potential was estimated by measuring fluorescence change of DiBAC in C6 glioma cells. Petatewalide B inhibited the antigen-induced degranulation of β-hexosaminidase in RBL-2H3 mast cells, but did not affect antigen-induced Ca(2+) increase in the cells. Petatewalide B also showed inhibition of the LPS-induced induction of iNOS, but not COX-2 in mouse peritoneal macrophages. Nitric oxide production was also inhibited by petatewalide B in macrophages. In the ovalbumin-induced asthma model, petatewalide B strongly inhibited accumulations of eosinophils, macrophages, and lymphocytes in bronchoalveolar lavage fluid. Petatewalide B increased the membrane potential of C6 glioma cells in a concentration-dependent manner. Petatewalide B from Petasites genus not only has anti-allergic and anti-inflammatory effects but also induces a transient increase of membrane potential in C6 glioma cells.
Sigma-Aldrich
Albumin, dinitrophenyl, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Giemsa Stain, Modified Solution, according to Giemsa