Merck
  • Protolichesterinic Acid, Isolated from the Lichen Cetraria islandica, Reduces LRRC8A Expression and Volume-Sensitive Release of Organic Osmolytes in Human Lung Epithelial Cancer Cells.
Protolichesterinic Acid, Isolated from the Lichen Cetraria islandica, Reduces LRRC8A Expression and Volume-Sensitive Release of Organic Osmolytes in Human Lung Epithelial Cancer Cells.
Phytotherapy research : PTR (2015-11-10)
Unnur Arna Thorsteinsdottir, Margret Thorsteinsdottir, Ian Henry Lambert
ABSTRACT
We have tested the effect of protolichesterinic acid (PA) on the activity of the volume-sensitive release pathway for the organic osmolyte taurine (VSOAC) and the expression of the leucine-rich-repeat-channel 8A (LRRC8A) protein, which constitutes an essential VSOAC component. Exposing human lung cancer cells (A549) to PA (20 µg/mL, 24 h) reduces LRRC8A protein expression by 25% and taurine release following osmotic cell swelling (320 → 200 mOsm) by 60%. C75 (20 µg/mL, 24 h), a γ-lactone with a C8 carbon fatty acid chain, reduces VSOAC activity by 30%, i.e. less than PA. Stearic acid (20 µg/mL, 24 h) has no effect on VSOAC. Hence, length of PA's fatty acid chain adds to γ-lactone's inhibitory action. 5-Lipoxygenase (5-LO) activity is essential for swelling-induced activation of VSOAC. PA has no effect on cellular concentration of leukotrienes (5-HETE/LTB4 ) under hypotonic conditions, excluding that PA mediated inhibition of VSOAC involves 5-LO inhibition. A549 cells exposed to the chemotherapeutic drug cisplatin (10 μM, 24 h) reveal signs of apoptosis, i.e. 25% reduction in cell viability as well as 1.3-, 1.5- and 3.3-fold increase in the expression of LRRC8A, Bax (regulator of apoptosis) and p21 (regulator of cell cycle progression), respectively. PA reduces cell viability by 30% but has no effect on p21/Bax expression. This excludes PA as a pro-apoptotic drug in A549 cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human LRRC8A
Sigma-Aldrich
4,4′-Diisothiocyanatostilbene-2,2′-disulfonic acid disodium salt hydrate, ≥80% (elemental analysis), powder
Sigma-Aldrich
Triton X-100, laboratory grade