- Effects of Fetal and Neonatal Murine Peripheral Blood Mononuclear Cells Infusion on MicroRNA-145 Expression in Renal Vascular Smooth Muscle Cells in MRL/lpr Mice.
Effects of Fetal and Neonatal Murine Peripheral Blood Mononuclear Cells Infusion on MicroRNA-145 Expression in Renal Vascular Smooth Muscle Cells in MRL/lpr Mice.
For patients with refractory systemic lupus erythematosus, current medications are insufficient to control their condition, and new treatments are necessary. We aimed to evaluate the therapeutic effect of fetal and neonatal murine peripheral blood (FNPB) mononuclear cells and their impact on microRNA-145 (miR-145) in renal vascular smooth muscle cells (VSMCs) of MRL/lpr lupus-prone mice. MRL/lpr mice aged 20 weeks were randomized to 3 groups of 15 (control group, radiation group, infusion group). The renal tissues were subjected to pathological examination. In situ hybridization assay was applied to measure miR-145 expression in renal vessels of MRL/lpr mice. The infusion group had significantly better results for pathological renal tissue lesions than either the control or radiation group. In MRL/lpr mice, there was positive expression of miR-145 in renal VSMCs, although the expression of miR-145 was not discernible in renal vascular intima and adventitia. The miR-145 expression in renal VSMCs in the infusion group was significantly higher than in the control or radiation group, and higher in the radiation group than in the control group; however, the difference was not statistically significant. The increased expression of miR-145 in renal VSMCs might be one of the mechanisms supporting FNPB as a therapy for lupus nephritis; it also suggests that the miR-145 in renal vessels might be a new target for treatment of lupus nephritis.