Merck
  • Design and synthesis of a potent inhibitor of class 1 DYRK kinases as a suppressor of adipogenesis.
Design and synthesis of a potent inhibitor of class 1 DYRK kinases as a suppressor of adipogenesis.
Bioorganic & medicinal chemistry (2015-07-07)
So Masaki, Isao Kii, Yuto Sumida, Tomoe Kato-Sumida, Yasushi Ogawa, Nobutoshi Ito, Mitsuhiro Nakamura, Rie Sonamoto, Naoyuki Kataoka, Takamitsu Hosoya, Masatoshi Hagiwara
ABSTRACT
Dysregulation of dual-specificity tyrosine-phosphorylation-regulated kinase 1A (DYRK1A) has been demonstrated in several pathological conditions, including Alzheimer's disease and cancer progression. It has been recently reported that a gain of function-mutation in the human DYRK1B gene exacerbates metabolic syndrome by enhancing obesity. In the previous study, we developed an inhibitor of DYRK family kinases (INDY) and demonstrated that INDY suppresses the pathological phenotypes induced by overexpression of DYRK1A or DYRK1B in cellular and animal models. In this study, we designed and synthesized a novel inhibitor of DYRK family kinases based on the crystal structure of the DYRK1A/INDY complex by replacing the phenol group of INDY with dibenzofuran to produce a derivative, named BINDY. This compound exhibited potent and selective inhibitory activity toward DYRK family kinases in an in vitro assay. Furthermore, treatment of 3T3-L1 pre-adipocytes with BINDY hampered adipogenesis by suppressing gene expression of the critical transcription factors PPARγ and C/EBPα. This study indicates the possibility of BINDY as a potential drug for metabolic syndrome.
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
SAFC
HEPES
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
1,3-Propylene sulfite, 99%
SAFC
HEPES
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications