Merck
Dissecting the roles of human BUB1 in the spindle assembly checkpoint.
Journal of cell science (2015-07-08)
Mathijs Vleugel, Tim A Hoek, Eelco Tromer, Tale Sliedrecht, Vincent Groenewold, Manja Omerzu, Geert J P L Kops
ABSTRACT
Mitotic chromosome segregation is initiated by the anaphase promoting complex/cyclosome (APC/C) and its co-activator CDC20 (forming APC/C(CDC20)). APC/C(CDC20) is inhibited by the spindle assembly checkpoint (SAC) when chromosomes have not attached to spindle microtubules. Unattached kinetochores catalyze the formation of a diffusible APC/C(CDC20) inhibitor that comprises BUBR1 (also known as BUB1B), BUB3, MAD2 (also known as MAD2L1) and a second molecule of CDC20. Recruitment of these proteins to the kinetochore, as well as SAC activation, rely on the mitotic kinase BUB1, but the molecular mechanism by which BUB1 accomplishes this in human cells is unknown. We show that kinetochore recruitment of BUBR1 and BUB3 by BUB1 is dispensable for SAC activation. Unlike its yeast and nematode orthologs, human BUB1 does not associate stably with the MAD2 activator MAD1 (also known as MAD1L1) and, although required for accelerating the loading of MAD1 onto kinetochores, BUB1 is dispensable for the maintenance of steady-state levels of MAD1 there. Instead, we identify a 50-amino-acid segment that harbors the recently reported ABBA motif close to a KEN box as being crucial for the role of BUB1 in SAC signaling. The presence of this segment correlates with SAC activity and efficient binding of CDC20 but not of MAD1 to kinetochores.
Sigma-Aldrich
MISSION® esiRNA, targeting human BUB1