Merck
Cortactin promotes colon cancer progression by regulating ERK pathway.
International journal of oncology (2015-07-08)
Qing-Feng Ni, Jia-Wei Yu, Fei Qian, Nai-Zhi Sun, Jian-Jia Xiao, Jian-Wei Zhu
ABSTRACT
Cortactin is upregulated in various cancers including breast cancer, head and neck squamous cell carcinoma and gastric cancer. However, the role of cortactin in the pathogenesis of colon cancer remains unclear. mRNA expression of cortactin in colon cancer samples and cell lines was detected by quantitative real-time PCR (qRT-PCR), while protein expression of cortactin in colon cancer tissues and adjacent non-cancer tissues was assessed by immunohistochemistry. The role of cortactin in regulation of the proliferation of colon cancer derived cells were investigated both in vitro and in vivo. In the total of 60 paired colon cancer specimens, compared with the adjacent non-cancer tissues, the expression of cortactin mRNA was upregulated in 45 (75.0%). Immunohistochemical analysis showed significantly increased cortactin expression in colon cancer (42/60, 70.0%) compared to control tissues (18/60, 30.0%). Overexpression of cortactin promoted HCT116 cellular colony formation and tumor growth. Conversely, cortactin knockdown inhibited these effects in SW480 cells. Mechanistic analyses indicated that cortactin was able to activate the EGFR-ERK signaling pathway. Additionally, cortactin expression was associated with tumor size, tumor stages and lymphatic invasion, increased cortactin expression predicts poor prognosis in patients with colon cancer. In summary, cortactin demonstrated the promotive effect in human colon cancer cell growth and tumorigenicity. These results indicated that cortactin may serve as an effective target for gene therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human CTTN
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cttn
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O