Merck
  • Megastigmane Glucosides and Megastigmanes from the Leaves of Meliosma lepidota ssp. squamulata.
Megastigmane Glucosides and Megastigmanes from the Leaves of Meliosma lepidota ssp. squamulata.
Chemical & pharmaceutical bulletin (2015-08-04)
Yuka Uemura, Mio Iwami, Susumu Kawakami, Sachiko Sugimoto, Katsuyoshi Matsunami, Hideaki Otsuka, Takakazu Shinzato, Masatoshi Kawahata, Kentaro Yamaguchi
ABSTRACT
From the leaves of Meliosma lepidota ssp. squamulata, megastigmane glucosides with spiro-structures and megastigmanes were isolated. Their structures were determined by X-ray crystallographic analyses and spectroscopic investigation. The absolute structures of the megastigmanes were determined by the modified Mosher's method.
Sigma-Aldrich
α-Methoxy-α-(trifluoromethyl)phenylacetic acid, 97%