Merck
  • Insecticide resistance and its underlying mechanisms in field populations of Aedes aegypti adults (Diptera: Culicidae) in Singapore.
Insecticide resistance and its underlying mechanisms in field populations of Aedes aegypti adults (Diptera: Culicidae) in Singapore.
Parasites & vectors (2014-10-11)
Sin-Ying Koou, Chee-Seng Chong, Indra Vythilingam, Chow-Yang Lee, Lee-Ching Ng
ABSTRACT
In Singapore, dose-response bioassays of Aedes aegypti (L.) adults have been conducted, but the mechanisms underlying resistance to insecticides remain unclear. In this study, we evaluated insecticide resistance and its underlying mechanism in field populations of Ae. aegypti adults. Seven populations of Ae. aegypti were collected from public residential areas and assays were conducted according to WHO guidelines to determine their susceptibility to several commonly used insecticides. Various levels of pyrethroid resistance (RR50 = 3.76 to 142.06-fold) and low levels of pirimiphos-methyl resistance (RR50 = 1.01 to 1.51-fold) were detected. The insecticide susceptibility profile of Ae. aegypti adults was homogenous among the different study sites. Addition of the synergists piperonyl butoxide, S,S,S,-tributyl phosphorotrithioate, and triphenyl phosphate generally failed to enhance the toxicity of the insecticides investigated, suggesting an insignificant role of metabolic-based insecticide resistance and possible involvement of target site resistance. Further biochemical investigation of specific metabolic enzyme activities provided further evidence that detoxifying enzymes such as mono-oxygenases, esterases, glutathione S-transferases and altered acethylcholinesterases generally did not contribute to the resistance observed. This study confirmed the presence of pyrethroid resistance among Ae. aegypti adults in Singapore and documented the early onset of organophosphate resistance.
Sigma-Aldrich
Triphenyl phosphate, ≥99%
Sigma-Aldrich
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D)
Sigma-Aldrich
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.