Merck
  • Telbivudine prevents vertical transmission of hepatitis B virus from women with high viral loads: a prospective long-term study.
Telbivudine prevents vertical transmission of hepatitis B virus from women with high viral loads: a prospective long-term study.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association (2014-09-25)
Quanxin Wu, Hongfei Huang, Xiaowen Sun, Meimin Pan, Yun He, Shun Tan, Yi Zeng, Li Li, Guohong Deng, Zehui Yan, Dengming He, Junnan Li, Yuming Wang
ABSTRACT
Hepatitis B virus (HBV) infection is a leading cause of liver diseases. We investigated the efficacy and safety of telbivudine in preventing transmission of HBV from hepatitis B e antigen-positive pregnant women with high viral loads to their infants in an open-label study. We performed a prospective study of 450 hepatitis B e antigen-positive pregnant women with HBV DNA levels greater than 10(6) IU/mL; 279 women received telbivudine (600 mg/d) during weeks 24 to 32 of gestation, and 171 women who were unwilling to take antiviral drugs participated as controls. All newborns were vaccinated with a recombinant HBV vaccine and hepatitis B immune globulin, according to a standard immunoprophylaxis procedure. Mother-to-child transmission of HBV was determined by detection of hepatitis B surface antigen and HBV DNA in the infant 6 months after birth. None of the infants whose mothers were given telbivudine tested positive for of hepatitis B surface antigen at 6 months, compared with 14.7% of infants in the control group (P = 5.317 × 10(-8)). Levels of HBV DNA also decreased among women given telbivudine; 40 of 172 (23.2%) women given telbivudine had undetectable HBV DNA levels before delivery, compared with none of the controls. A significantly higher proportion of women given telbivudine had undetectable levels of HBV DNA in cord blood (99.1%) than controls (61.5%; P = 1.195 × 10(-22)). No severe adverse events or complications were observed in women or infants. Telbivudine significantly reduces vertical transmission of HBV from pregnant women to their infants; it is safe and well tolerated by women and infants. Antiretroviral Pregnancy Registry Health Care Providers ID: 26592; Government number: Natural Science Foundation of China (NSFC) 30830090, 30972598; and Third Military Medical University Key Project for Clinical Research: 2012XLC05).
Sigma-Aldrich
Telbivudine, ≥98% (HPLC)