Merck
  • Overexpression of lncRNA H19 enhances carcinogenesis and metastasis of gastric cancer.
Overexpression of lncRNA H19 enhances carcinogenesis and metastasis of gastric cancer.
Oncotarget (2014-05-09)
Hao Li, Beiqin Yu, Jianfang Li, Liping Su, Min Yan, Zhenggang Zhu, Bingya Liu
ABSTRACT
Long non-coding RNAs (lncRNAs) play key roles in the progression and metastasis of some carcinomas. We previously showed that the expression of lncRNA H19 (H19) was higher in gastric cancer (GC) tissues than that in paired noncanerous tissues. However, the underlying mechanisms remain unclear. In this study, H19/miR-675 knockdown models in the MKN45 cell line and ectopic expression models in the SGC7901 cell line were established, and a co-expression network of H19 was generated to identify target genes by RIP and DLR. The results showed that overexpression of H19 promoted the features of GC including proliferation, migration, invasion and metastasis. An H19 co-expression network identified ISM1 as a binding protein of H19, and its expression was positively correlated with that of H19. CALN1 was identified as a target gene of miR-675 and its expression was negatively correlated with that of miR-675. H19 and MiR-675 function in a similar manner. However, H19 RNA actively binds to ISM1 and miR-675 targets CALN1. These differences suggest that H19 plays other roles besides encoding miR-675 in GC. Our results suggest that the effect of H19 in GC is mediated by the direct upregulation of ISM1 and the indirect suppression of CALN1 expression via miR-675.
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.