Merck
  • Targeted transplantation of human umbilical cord blood endothelial progenitor cells with immunomagnetic nanoparticles to repair corneal endothelium defect.
Targeted transplantation of human umbilical cord blood endothelial progenitor cells with immunomagnetic nanoparticles to repair corneal endothelium defect.
Stem cells and development (2014-10-16)
Chunyi Shao, Junzhao Chen, Ping Chen, Mengyu Zhu, Qinke Yao, Ping Gu, Yao Fu, Xianqun Fan
ABSTRACT
Corneal endothelial dysfunction involves progressive corneal edema and loss of visual acuity, which result in the need for corneal transplantation. The global shortage of donor corneas limits the development of the surgery. Reconstruction of a bioengineered corneal endothelium might resolve this problem. Various scaffolds have been used, but poor biocompatibility and degradation limit their applications. In this study, a novel method of targeted cellular transplantation without permanent residence of cell carriers in the host was proposed. Human umbilical cord blood endothelial progenitor cells (UCB EPCs) were labeled with CD34 immunomagnetic nanoparticles. The efficiency of the magnet attraction was evaluated in vitro with a simple device simulating the anterior chamber. The UCB EPCs labeled with nanoparticles were transplanted into the anterior chamber of rabbits with magnet attraction. The results indicated that labeling the nanoparticles did not affect the proliferation of the UCB EPCs. The in vitro study indicated that the magnet could directionally attract UCB EPCs labeled with nanoparticles. The in vivo study indicated that the corneas in rabbits transplanted with UCB EPCs labeled with nanoparticles and magnet attraction became relatively transparent with little edema. These results showed that UCB EPCs labeled with CD34 immunomagnetic nanoparticles could be attracted directionally by a magnet and could repair corneal endothelial defects, providing a promising cell therapy for corneal endothelial dysfunction.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium DL-lactate solution, syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Hematoxylin
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material