Merck
  • Improving selectivity preserving affinity: new piperidine-4-carboxamide derivatives as effective sigma-1-ligands.
Improving selectivity preserving affinity: new piperidine-4-carboxamide derivatives as effective sigma-1-ligands.
European journal of medicinal chemistry (2014-12-22)
Daniele Zampieri, Erik Laurini, Luciano Vio, Maurizio Fermeglia, Sabrina Pricl, Bernhard Wünsch, Dirk Schepmann, Maria Grazia Mamolo
ABSTRACT
We report the design, synthesis and binding evaluation against σ1 and σ2 receptors of a series of new piperidine-4-carboxamide derivatives variously substituted on the amide nitrogen atom. Specifically, we assessed the effects exerted on σ receptor affinity by substituting the N-benzylcarboxamide group present on a series of compounds previously synthesized in our laboratory with different cyclic or linear moieties. The synthesized compounds 2a-o were tested to estimate their affinity and selectivity toward σ1 and σ2 receptors. Very high σ1 affinity (Ki = 3.7 nM) and Kiσ2/Kiσ1 selectivity ratio (351) were found for the tetrahydroquinoline derivative 2k, featuring a 4-chlorobenzyl moiety linked to the piperidine nitrogen atom.
Sigma-Aldrich
4-Chloro-DL-phenylalanine
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
(−)-Pentazocine, ≥98% (HPLC)
Sigma-Aldrich
(+)-Pentazocine, solid