Merck
  • Proteomic analysis of differentially expressed proteins in 5-fluorouracil-treated human breast cancer MCF-7 cells.
Proteomic analysis of differentially expressed proteins in 5-fluorouracil-treated human breast cancer MCF-7 cells.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico (2013-11-13)
J Cai, S Chen, W Zhang, Y Wei, J Lu, J Xing, Y Dong
ABSTRACT
5-Fluorouracil (5-Fu) is a commonly used chemotherapeutic agent in clinical care of breast cancer patients. However, the mechanism of how the 5-Fu works is complex and still largely unknown. The objective of this study was to understand the mechanism further and explore the new targets of 5-Fu. The differentially expressed proteins induced by 5-Fu in human breast cancer MCF-7 cells were identified by proteomic analysis. Four differentially expressed proteins were validated using Western blot and quantitative real-time reverse-transcription polymerase chain reaction analysis for protein and mRNA levels. The effect of 5-Fu on MCF-7 cells was determined by cell viability assay, transmission electron microscopy and flow cytometry analysis. 5-Fu dose-dependently inhibited cell proliferation with the IC50 value of 98.2 μM. 5-Fu also induced obviously morphological change and apoptosis in MCF-7 cells. Twelve differentially expressed proteins involved in energy metabolism, cytoskeleton, cellular signal transduction and tumor invasion and metastasis were identified. These results may provide a new insight into the molecular mechanism of 5-Fu in therapy of breast cancer.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester, BioReagent, suitable for fluorescence, mixture of isomers, ≥80% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Propidium iodide solution