Merck
  • Drug effects on multiple and concurrent schedules of ethanol- and food-maintained behaviour: context-dependent selectivity.
Drug effects on multiple and concurrent schedules of ethanol- and food-maintained behaviour: context-dependent selectivity.
British journal of pharmacology (2014-04-05)
B C Ginsburg, R J Lamb
ABSTRACT
Drugs that more potently or effectively reduce ethanol-maintained behaviour versus an alternative are considered selective and are considered promising pharmacotherapies for alcoholism. Such results are often obtained using separate groups or multiple schedules where ethanol and the alternative are available alone or sequentially. Recently, we observed that when ethanol and food were available sequentially under a multiple schedule, fluvoxamine and varenicline were selective; yet this selectivity disappeared when ethanol and food were concurrently available. We examined the generality of these findings by comparing doses of several drugs required to decrease ethanol- and food-maintained responding under a multiple schedule and under a concurrent schedule. Effects were determined for chlordiazepoxide, 2,5-dimethoxy-4-iodoamphetamine (DOI), meta-chlorophenylpiperazine (mCPP), morphine, naltrexone and d-amphetamine. Under the multiple schedule, ED50 values for decreases in ethanol-maintained responding were significantly different and lower than ED50 s for decreases in food-maintained responding (demonstrating selectivity) for each drug except for chlordiazepoxide (which was equipotent) and naltrexone (which did not affect responding). However, this selectivity vanished or even inverted under the concurrent schedule, such that ED50 values for decreasing ethanol- and food-maintained responding were not different (or, following DOI, the ED50 for food-maintained responding was lower than for ethanol-maintained responding). Results are consistent with those seen following fluvoxamine and varenicline administration, and suggest that selectivity is assay-dependent. These results indicate the need for careful interpretation of selective drug effects, especially when obtained in situations where ethanol or the alternative is the only programmed reinforcement available.
Product Description
Chlordiazepoxide hydrochloride, European Pharmacopoeia (EP) Reference Standard