Merck
  • Evaluation of the effect of intra-operative intravenous fluid on post-operative pain and pulmonary function: a randomized trial comparing 10 and 30 ml kg(-1) of crystalloid.
Evaluation of the effect of intra-operative intravenous fluid on post-operative pain and pulmonary function: a randomized trial comparing 10 and 30 ml kg(-1) of crystalloid.
Irish journal of medical science (2013-12-11)
B D Straub, A Aslani, K Enohumah, R Rahore, I Conrick-Martin, D Kumar, M Campbell, P Dicker, E Mocanu, J P Loughrey, N E Hayes, C L McCaul
ABSTRACT
Existing evidence suggests that administration of intravenous fluids has been shown to improve outcomes including pain in gynecological laparoscopic surgery but the optimum fluid dose has not been determined. To determine the effect of administration of intravenous fluids on post-operative pain and pulmonary function after gynecological laparoscopy. In a prospective randomized double-blinded study 100 ASA 1 and 2 elective patients undergoing gynecological laparoscopy were randomized to receive intravenous compound sodium lactate 10 ml kg(-1) (CSL10-restrictive) or 30 ml kg(-1) (CSL30-liberal) administered intra-operatively. The primary outcome measure was the post-operative pain score at 24, 48 and 72 h, assessed by 0-10 verbal rating scale (VRS). Pulmonary function (FEV1, FVC, PEFR) and oxygen saturation were also measured. Patients who received CSL 30 had lower post-operative pain scores than CSL 10 (ANCOVA-mean difference = 0.47, 95 % CI 0.11-0.83, P = 0.01). Post-operative pain VRS was lower in CSL30 than CSL10 at 48 h (mean difference 0.56, 95 % CI 0.04-1.09, P = 0.036). Patients in CSL30 reported shoulder tip pain less frequently than those in CSL10 (30.4 vs. 43.9 % of assessments, P = 0.03, OR 0.58) but reported wound pain more frequently 39.0 vs. 24.2 %, P = 0.01, OR 2.0). Indices of pulmonary function did not differ between groups at any time. Liberal compared to restrictive administration of i.v. crystalloid is associated with a clinical modest reduction in pain. Pulmonary dysfunction was not increased with liberal fluid administration.
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)