Merck
  • Nociceptin/orphanin FQ slows inspiratory rhythm via its direct effects on the pre-Bötzinger complex.
Nociceptin/orphanin FQ slows inspiratory rhythm via its direct effects on the pre-Bötzinger complex.
Respiratory physiology & neurobiology (2014-12-17)
Koichi Takita, Yuji Morimoto
ABSTRACT
In a previous study, we showed that in an in vitro en bloc preparation of newborn rats perfused with standard [K(+)] (6.2mM) and high [K(+)] (11.2mM) artificial cerebrospinal fluid (aCSF), nociceptin/orphanin FQ (N/OFQ) suppresses bursting of pre-inspiratory neurons with 1:1 coupling to the fictive inspiration. However, it is unclear whether the pre-Bötzinger complex (preBötC) is involved in the N/OFQ-induced slowing. Using in vitro en bloc preparations with and without the retrotrapezoid nucleus/parafacial respiratory group (RTN/pFRG) perfused with high [K(+)] aCSF, we found the following: (1) there were no differences in the effects of N/OFQ on the inspiratory rhythm between the preparations with and without the RTN/pFRG, (2) N/OFQ decreased the input resistance of inspiratory neurons (Insps) in the preparations without the RTN/pFRG and suppressed their ectopic firing activities, and (3) N/OFQ suppressed the spontaneous firing of Insps under a chemical synaptic transmission blockade. In conclusion, it is possible that the preBötC is involved in N/OFQ-induced inspiratory rhythm slowing.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Caffeine, European Pharmacopoeia (EP) Reference Standard
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard