Merck
  • Comparison of shockwave lithotripsy and flexible ureteroscopy for the treatment of kidney stones in patients with a solitary kidney.
Comparison of shockwave lithotripsy and flexible ureteroscopy for the treatment of kidney stones in patients with a solitary kidney.
Journal of endourology (2014-10-01)
Emrah Yuruk, Murat Binbay, Faruk Ozgor, Levent Sekerel, Yalcin Berberoglu, Ahmet Yaser Muslumanoglu
ABSTRACT
To compare the outcomes of these minimally invasive procedures in this patient population. The database of our institution has been retrospectively reviewed, and medical records of urolithiasis patients with a solitary kidney who underwent flexible ureteroscopy (F-URS) or extracorporeal shock wave lithotripsy (SWL) between January 2009 and December 2012 were examined. Retreatment rates, complications, changes in estimated glomerular filtration rates (eGFRs), chronic kidney disease (CKD) stages, and stone-free rates were compared between the two groups. Stones of 48 patients (mean age: 48.8±15.4, range: 14-76) with solitary kidneys were treated with SWL (n=30, 62.5%) or F-URS (n=18, 37.5%). Patient demographics and stone related parameters were similar. The most common stone location was the pelvis in the SWL group (36.6%), whereas it was the pelvis and a calix in the F-URS group (38.8%). Complications and success rates were similar in both groups, however, patients in the SWL group needed more sessions to achieve stone clearance (2.2±0.89 vs 1.06±0.24, p=0.0001). Preoperative and postoperative eGFR and CKD stage changes were also similar. Both SWL and F-URS are effective and safe techniques, which can be used for the treatment of stones in patients with solitary kidneys. However, patients treated with SWL need more sessions to achieve stone clearance.
Product Description
Tramadol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tramadol hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar