Merck
  • Characterisation of a new human and murine member of the DnaJ family of proteins.
Characterisation of a new human and murine member of the DnaJ family of proteins.
Biochemical and biophysical research communications (1998-02-25)
G Silins, S Grimmond, N Hayward
ABSTRACT
We report the characterisation of a human gene, designated MCG18 (multiple endocrine neoplasia type 1 candidate gene 18), that encodes a new member of the DnaJ family of proteins. Database searches indicate that MCG18 also has the locus name HSPF2. MCG18 lies 250bp centromeric of the VRF/VEGFB gene on chromosome 11q13. The MCG18 cDNA is predicted to encode a 241 amino acid product that has partial homology to Escherichia coli dnaJ in that it contains the J domain. However, MCG18 has greatest similarity to a functionally undefined protein from Caenorhabditis elegans, both of which are predicted to have a membrane-spanning region adjacent to their J domains. The cDNA encoding the murine homolog (Mcg18) was also cloned and sequenced, and the encoded protein shares 81% similarity to MCG18. The coding region of MCG18 is interrupted by 4 introns and the mRNA is expressed as a 1.4kb message in all tissues examined, including those derived from the breast, ovary, bladder, lung and keratinocytes.