Merck
  • Tandem thia-Fries rearrangement--cyclisation of 2-(trimethylsilyl)phenyl trifluoromethanesulfonate benzyne precursors.
Tandem thia-Fries rearrangement--cyclisation of 2-(trimethylsilyl)phenyl trifluoromethanesulfonate benzyne precursors.
Chemical communications (Cambridge, England) (2013-07-24)
Catherine Hall, Jaclyn L Henderson, Guillaume Ernouf, Michael F Greaney
ABSTRACT
A novel transformation of 2-(trimethylsilyl)phenyl trifluoromethanesulfonate aryne precursors is described.
Sigma-Aldrich
2-(Trimethylsilyl)phenyl trifluoromethanesulfonate, 97%