Merck
  • A role for mammalian Ubc6 homologues in ER-associated protein degradation.
A role for mammalian Ubc6 homologues in ER-associated protein degradation.
Journal of cell science (2002-06-26)
Uwe Lenk, Helen Yu, Jan Walter, Marina S Gelman, Enno Hartmann, Ron R Kopito, Thomas Sommer
ABSTRACT
Integral membrane and secretory proteins which fail to fold productively are retained in the endoplasmic reticulum and targeted for degradation by cytoplasmic proteasomes. Genetic and biochemical analyses suggest that substrates of this pathway must be dislocated across the membrane of the endoplasmic reticulum (ER) by a process requiring a functional Sec61 complex and multiubiquitinylation. In yeast, the tail-anchored ubiquitin-conjugating enzyme Ubc6p, which is localized to the cytoplasmic surface of the ER, participates in ER-associated degradation (ERAD) of misfolded proteins. Here we describe the identification of two families of mammalian Ubc6p-related proteins. Members of both families are also located in the ER membrane and display a similar membrane topology as the yeast enzyme. Furthermore we show that expression of elevated levels of wild-type and dominant-negative alleles of these components affects specifically ERAD of the alpha subunit of the T-cell receptor and a mutant form of the CFTR protein. Similarly, we describe that the expression level of Ubc6p in yeast is also critical for ERAD, suggesting that the Ubc6p function is highly conserved from yeast to mammals.