Merck
  • Enantioselective syntheses of syn- and anti-β-hydroxyallylsilanes via allene hydroboration-aldehyde allylboration reactions.
Enantioselective syntheses of syn- and anti-β-hydroxyallylsilanes via allene hydroboration-aldehyde allylboration reactions.
Organic letters (2011-03-18)
Ming Chen, William R Roush
ABSTRACT
The kinetic hydroboration of allenylsilane 5 with ((d)Ipc)(2)BH at -40 °C provides allylborane 9Z with ≥12:1 selectivity. When the hydroboration is performed at temperatures above -40 °C, 9Z isomerizes to the thermodynamically more stable allylborane 9E with >20:1 selectivity. Subsequent treatment of 9Z or 9E with aldehydes at -78 °C provides syn- or anti-β-hydroxyallylsilanes, 7 or 8, respectively.
Sigma-Aldrich
Hydrocinnamaldehyde, technical grade, 90%