Merck
Late-stage deoxyfluorination of alcohols with PhenoFluor.
Journal of the American Chemical Society (2013-02-13)
Filippo Sladojevich, Sophie I Arlow, Pingping Tang, Tobias Ritter
ABSTRACT
An operationally simple protocol for the selective deoxyfluorination of structurally complex alcohols is presented. Several fluorinated derivatives of natural products and pharmaceuticals have been prepared to showcase the potential of the method for late-stage diversification and its functional group compatibility. A series of simple guidelines for predicting the selectivity in substrates with multiple alcohols is given.
Sigma-Aldrich
Phenofluor solution, 0.1 M in toluene
Pricing and availability is not currently available.