Merck
  • Latexin is down-regulated in hematopoietic malignancies and restoration of expression inhibits lymphoma growth.
Latexin is down-regulated in hematopoietic malignancies and restoration of expression inhibits lymphoma growth.
PloS one (2012-10-03)
Yi Liu, Dianna Howard, Kyle Rector, Carol Swiderski, Jason Brandon, Lawrence Schook, Jayesh Mehta, J Scott Bryson, Subbarao Bondada, Ying Liang
ABSTRACT
Latexin is a negative regulator of hematopoietic stem cell number in mice. Its dysregulated expression in other tumors led us to hypothesize that latexin may have tumor suppressor properties in hematological malignancies. We found that latexin was down-regulated in a variety of leukemia and lymphoma cell lines as well as in CD34+ cells from the blood and marrow of patients with these malignancies. 5-aza-2'-deoxycytodine treatment and bisulfite sequencing revealed hypermethylation of latexin promoter in tumor cells. Retrovirus-mediated latexin overexpression in A20 mouse lymphoma cells inhibited their in vitro growth by 16 fold and in vivo tumor volume by 2 fold. Latexin caused growth inhibition of lymphoma cells by significantly increasing apoptosis through the down-regulation of anti-apoptotic genes Bcl-2 and Pim-2. The molecular mechanism underlying latexin-mediated tumor inhibition was not through its canonical carboxypeptidase inhibitor activity. These results are consistent with a tumor suppressor role for latexin and suggest that latexin may have clinical efficacy in the treatment of malignancies.