Merck
  • Comparative study on the derivatization of l-alpha-acetyl-methadol metabolites for electron capture gas-liquid chromatography.
Comparative study on the derivatization of l-alpha-acetyl-methadol metabolites for electron capture gas-liquid chromatography.
Journal of chromatography (1976-12-22)
D H Lau, G L Henderson
ABSTRACT
Six reagents-trichloroacetyl chloride, trichloroacetic anhydride, pentafluorobenzoyl chloride, heptafluorobutyryl chloride, heptafluorobutyric anhydride, and trifluoroacetic anhydride- were evaluated as potential derivatizing reagents for quantitating the metabolites of l-alpha-acetylmethadol (LAAM) -noracetylmethadol, dinoracetylmethadol, methadol, and normethadol-by electron capture gas-liquid chromatography. All of the reagents studied reacted quantitatively with all of the metabolites except methadol; however, trichloroacetyl chloride was found to be the most satisfactory general reagent for analyzing these metabolites in biological fluids. A gas-liquid chromatographic method is presented which combines both flame ionization and electron capture detection for quantitating plasma and urine levels of methadone, l-alpha-acetylmethadol and its metabolites.
Sigma-Aldrich
Trichloroacetic anhydride, technical grade, 95%
Pricing and availability is not currently available.