Merck
  • Metachelins, mannosylated and N-oxidized coprogen-type siderophores from Metarhizium robertsii.
Metachelins, mannosylated and N-oxidized coprogen-type siderophores from Metarhizium robertsii.
Journal of natural products (2014-07-06)
Stuart B Krasnoff, Ivan Keresztes, Bruno G G Donzelli, Donna M Gibson
ABSTRACT
Under iron-depleted culture conditions, the entomopathogenic fungus Metarhizium robertsii (Bischoff, Humber, and Rehner) (= M. anisopliae) produces a complex of extracellular siderophores including novel O-glycosylated and N-oxidized coprogen-type compounds as well as the known fungal siderophores N(α)-dimethylcoprogen (NADC) and dimerumic acid (DA). Metachelin A (1), the most abundant component in the M. robertsii siderophore mixture, was characterized as a 1094 Da analogue of NADC that is O-glycosylated by β-mannose at both terminal hydroxyl groups and N-oxidized at the dimethylated α-nitrogen. The mixture also contained a 1078 Da analogue, metachelin B (2), which lacks the N-oxide modification. Also characterized were the aglycone of 1, i.e., the N-oxide of NADC (3), and the monomannoside of DA (6). N-Oxide and O-glycosyl substituents are unprecedented among microbial siderophores. At high ESIMS source energy and at room temperature in DMSO, 1 underwent Cope elimination, resulting in loss of the N(α)-dimethyl group and dehydration of the α-β bond. High-resolution ESIMS data confirmed that all tri- and dihydroxamate siderophores (1-6) complex with trivalent Fe, Al, and Ga. In a chrome azurol S assay, all of the M. robertsii siderophores showed iron-binding activity roughly equivalent to that of desferrioxamine B.
Product Description
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Iron, foil, 6mm disks, thickness 0.004mm, 99.85%
Iron, foil, 50mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.004mm, 99.99+%
Iron, foil, 8mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, as rolled, 99.99+%
Iron, foil, light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Iron, foil, light tested, 50x50mm, thickness 0.025mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.005mm, 99.85%
Iron, foil, 50mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.020mm, 99.85%
Iron, foil, 5m coil, thickness 0.1mm, hard, 99.5%
Iron, foil, light tested, 50x50mm, thickness 0.038mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, 6mm disks, thickness 0.15mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 8mm disks, thickness 0.003mm, 99.85%
Iron, foil, 5m coil, thickness 0.20mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.008mm, 99.85%
Iron, foil, 5m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.008mm, 99.85%