Merck
  • Dexamethasone for the prevention of postpericardiotomy syndrome: A DExamethasone for Cardiac Surgery substudy.
Dexamethasone for the prevention of postpericardiotomy syndrome: A DExamethasone for Cardiac Surgery substudy.
American heart journal (2014-06-24)
Jeroen J H Bunge, Dirk van Osch, Jan M Dieleman, Kirolos A Jacob, Jolanda Kluin, Diederik van Dijk, Hendrik M Nathoe
ABSTRACT
The postpericardiotomy syndrome (PPS) is a common complication following cardiac surgery. The pathophysiology remains unclear, although evidence exists that surgical trauma and the use of cardiopulmonary bypass provoke an immune response leading to PPS. We hypothesized that an intraoperative dose of dexamethasone decreases the risk of PPS, by reducing this inflammatory response. We performed a subanalysis of the DECS study, which is a multicenter, double-blind, placebo-controlled, randomized trial of 4,494 patients undergoing cardiac surgery with use of cardiopulmonary bypass. The aim of the DECS study was to investigate whether a single intraoperative dose of 1 mg/kg dexamethasone reduced the incidence of a composite of death, myocardial infarction, stroke, renal failure, or respiratory failure, within 30 days of randomization. In this substudy, we retrospectively analyzed the occurrence of PPS in 822 patients who were included in the DECS trial and underwent valvular surgery. Postpericardiotomy syndrome was diagnosed if 2 of 5 listed symptoms were present: unexplained fever, pleuritic chest pain, pericardial or pleural rub, new or worsening pericardial or pleural effusion. All medical charts, x-rays, and echocardiograms were reviewed. Secondary end point was the occurrence of complicated PPS, defined as PPS with need for evacuation of pleural effusion, pericardiocentesis, and tamponade requiring intervention or hospital readmission for PPS. This is a blinded, single-center, post hoc analysis. Postpericardiotomy syndrome occurred in 119 patients (14.5%). The incidence of PPS after dexamethasone compared with placebo was 13.5% vs 15.5% (relative risk 0.88, 95% CI 0.63-1.22). For complicated PPS, the incidence was 3.8% versus 3.2% (relative risk 1.17, 95% CI 0.57-2.41, P = .66), respectively. In patients undergoing valvular cardiac surgery, high-dose dexamethasone treatment had no protective effect on the occurrence of PPS or complicated PPS.
Product Description
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard