Merck
  • H1-antihistamines exacerbate high-fat diet-induced hepatic steatosis in wild-type but not in apolipoprotein E knockout mice.
H1-antihistamines exacerbate high-fat diet-induced hepatic steatosis in wild-type but not in apolipoprotein E knockout mice.
American journal of physiology. Gastrointestinal and liver physiology (2014-05-24)
Vineesh V Raveendran, Karen M Kassel, Donald D Smith, James P Luyendyk, Kurt J Williams, Rachel Cherian, Gregory A Reed, Colleen A Flynn, Iván L Csanaky, Andrew L Lickteig, Matthew J Pratt-Hyatt, Curtis D Klaassen, Kottarappat N Dileepan
ABSTRACT
We examined the effects of two over-the-counter H1-antihistamines on the progression of fatty liver disease in male C57Bl/6 wild-type and apolipoprotein E (ApoE)-/- mice. Mice were fed a high-fat diet (HFD) for 3 mo, together with administration of either cetirizine (4 mg/kg body wt) or fexofenadine (40 mg/kg body wt) in drinking water. Antihistamine treatments increased body weight gain, gonadal fat deposition, liver weight, and hepatic steatosis in wild-type mice but not in ApoE-/- mice. Lobular inflammation, acute inflammation, and necrosis were not affected by H1-antihistamines in either genotype. Serum biomarkers of liver injury tended to increase in antihistamine-treated wild-type mice. Serum level of glucose was increased by fexofenadine, whereas lipase was increased by cetirizine. H1-antihistamines reduced the mRNA expression of ApoE and carbohydrate response element-binding protein in wild-type mice, without altering the mRNA expression of sterol regulatory element-binding protein 1c, fatty acid synthase, or ApoB100, in either genotype. Fexofenadine increased both triglycerides and cholesterol ester, whereas cetirizine increased only cholesterol ester in liver, with a concomitant decrease in serum triglycerides by both antihistamines in wild-type mice. Antihistamines increased hepatic levels of conjugated bile acids in wild-type mice, with the effect being significant in fexofenadine-treated animals. The increase was associated with changes in the expression of organic anion transport polypeptide 1b2 and bile salt export pump. These results suggest that H1-antihistamines increase the progression of fatty liver disease in wild-type mice, and there seems to be an association between the severity of disease, presence of ApoE, and increase in hepatic bile acid levels.
Sigma-Aldrich
Cetirizine dihydrochloride, ≥98% (HPLC)
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Cetirizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cetirizin dihydrochloride, ≥98.0% (HPLC)
Sigma-Aldrich
Terfenadine