Merck
  • Discovery and investigation of a novel class of thiophene-derived antagonists of the human glucagon receptor.
Discovery and investigation of a novel class of thiophene-derived antagonists of the human glucagon receptor.
Bioorganic & medicinal chemistry letters (2005-02-17)
Joseph L Duffy, Brian A Kirk, Zenon Konteatis, Elizabeth L Campbell, Rui Liang, Edward J Brady, Mari Rios Candelore, Victor D H Ding, Guoqiang Jiang, Frank Liu, Sajjad A Qureshi, Richard Saperstein, Deborah Szalkowski, Sharon Tong, Lauri M Tota, Dan Xie, Xiaodong Yang, Peter Zafian, Song Zheng, Kevin T Chapman, Bei B Zhang, James R Tata
ABSTRACT
A novel class of antagonists of the human glucagon receptor (hGCGR) has been discovered. Systematic modification of the lead compound identified substituents that were essential for activity and those that were amenable to further optimization. This SAR exploration resulted in the synthesis of 13, which exhibited good potency as an hGCGR functional antagonist (IC50 = 34 nM) and moderate bioavailability (36% in mice).
Sigma-Aldrich
2-Ethylbutyryl chloride, 97%
Pricing and availability is not currently available.