Merck
Methemoglobinemia caused by benzocaine topical spray.
Southern medical journal (1997-09-26)
H A Cooper
ABSTRACT
Acute methemoglobinemia is a medical emergency that can rapidly become fatal. A substantial number of drugs, including topical preparations, can precipitate this condition. I report a rare example of severe, acute methemoglobinemia caused by oropharyngeal anesthesia with topical benzocaine spray for orogastric intubation. The pathophysiology, diagnosis, and treatment of methemoglobinemia are reviewed briefly.
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Pricing and availability is not currently available.
Benzocaine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Pricing and availability is not currently available.