Merck
  • All-polyethylene tibial components are equal to metal-backed components: systematic review and meta-regression.
All-polyethylene tibial components are equal to metal-backed components: systematic review and meta-regression.
Clinical orthopaedics and related research (2012-09-14)
Klaas Auke Nouta, Wiebe C Verra, Bart G Pijls, Jan W Schoones, Rob G H H Nelissen
ABSTRACT
Less than 1% of all primary TKAs are performed with an all-polyethylene tibial component, although recent studies indicate all-polyethylene tibial components are equal to or better than metal-backed ones. We asked whether the metal-backed tibial component was clinically superior to the all-polyethylene tibial component in primary TKAs regarding revision rates and clinical functioning, and which modifying variables affected the revision rate. We systematically reviewed the literature for clinical studies comparing all-polyethylene and metal-backed tibial components used in primary TKAs in terms of revision rates, clinical scores, and radiologic parameters including radiostereometric analysis (RSA). Meta-regression techniques were used to explore factors modifying the observed effect. Our search yielded 1557 unique references of which 26 articles were included, comprising more than 12,500 TKAs with 231 revisions for any reason. Meta-analysis showed no differences between the all-polyethylene and metal-backed components except for higher migration of the metal-backed components. Meta-regression showed strong evidence that the all-polyethylene design has improved with time compared with the metal-backed design. The all-polyethylene components were equivalent to metal-backed components regarding revision rates and clinical scores. The all-polyethylene components had better fixation (RSA) than the metal-backed components. The belief that metal-backed components are better than all-polyethylene ones seems to be based on studies from earlier TKAs. This might no longer be true for modern TKAs. Level II, therapeutic study. See Guidelines for Authors for a complete description of levels of evidence.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Polyethylene (LDPE), ERM®, certified reference material