Merck
  • [Liposomal amphotericin B: a unique pharmacokinetic profile. An unfinished story].
[Liposomal amphotericin B: a unique pharmacokinetic profile. An unfinished story].
Revista espanola de quimioterapia : publicacion oficial de la Sociedad Espanola de Quimioterapia (2012-04-11)
José Ramón Azanza Perea, José Barberán
ABSTRACT
Amphotericin B in its lipid formulation continues to be the reference drug in the treatment of systemic fungal infections despite the time elapse since the development of this compound. The absence of fungal resistance, pharmacokinetics, and the better tolerability profile as compared with the remaining formulations of amphotericin B are sufficient reasons to justify its prominent therapeutic role. The liposome containing liposomal amphotericin B is very stable in relation to the presence of cholesterol and phospholipids are not thermolabile, so that free amphotericin B is almost inexistent (<1%), which explains the reduced incidence of effects related to the drug administration, and a reduction in the incidence of nephrotoxicity (half than that with amphotericin B lipid complex) and that even in some studies at doses of 1 mg/kg has been shown to be negligible. This profile explains the very high plasma drug concentrations and the reduced distribution volume and clearance, with a very prolonged elimination half-life. There are evidences showing that the liposome through amphotericin B is capable of binding to ergosterol present in the fungal membrane and only at this moment would be the antifungal released to exert its pharmacological effects.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Amphotericin B for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard