Merck
  • Survivin-dependent and -independent pathways and the induction of cancer cell death by tetra-O-methyl nordihydroguaiaretic acid.
Survivin-dependent and -independent pathways and the induction of cancer cell death by tetra-O-methyl nordihydroguaiaretic acid.
Seminars in oncology (2006-08-08)
Ru Chih C Huang, Chih Chuan Chang, David Mold
ABSTRACT
The inhibitor of apoptosis protein (IAP) family encodes a group of Baculovirus IAP repeat domain (BIR)-containing proteins that suppress apoptosis. Some of the IAPs, survivin and XIAP in particular, are differentially overexpressed in many types of human cancer and are deemed attractive anticancer targets. Here we review the regulation of survivin expression and survivin's functions in both normal and cancerous cells, and some of the current survivin-targeted cancer therapy. We further discuss the possible mechanisms of tetra-O-methyl nordihydroguaretic acid (M(4)N), a global transcription inhibitor, in the induction of cancer cell death in tumors via survivin-dependent and -independent pathways.
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥90% (HPLC), from Larrea divaricata (creosote bush)