Merck
Vinyl chloride: still a cause for concern.
Environmental health perspectives (2000-07-25)
J Kielhorn, C Melber, U Wahnschaffe, A Aitio, I Mangelsdorf
ABSTRACT
Vinyl chloride (VC) is both a known carcinogen and a regulated chemical, and its production capacity has almost doubled over the last 20 years, currently 27 million tons/year worldwide. According to recent reports it is still a cause for concern. VC has been found as a degradation product of chloroethylene solvents (perchloroethylene and trichloroethylene) and in landfill gas and groundwater at concentrations up to 200 mg/m(3) and 10 mg/L, respectively. Worldwide occupational exposure to VC still seems to be high in some countries (e.g., averages of approximately 1,300 mg/m(3) until 1987 in one factory), and exposure may also be high in others where VC is not regulated. By combining the most relevant epidemiologic studies from several countries, we observed a 5-fold excess of liver cancer, primarily because of a 45-fold excess risk from angiosarcoma of the liver (ASL). The number of ASL cases reported up to the end of 1998 was 197 worldwide. The average latency for ASL is 22 years. Some studies show a small excess risk for hepatocellular carcinoma, and others suggest a possible risk of brain tumors among highly exposed workers. Lung cancer, lymphomas, or leukemia do not seem to be related to VC exposure according to recent results. The mutation spectra observed in rat and human liver tumors (ASL and/or hepatocellular carcinoma) that are associated with exposure to VC are clearly distinct from those observed in sporadic liver tumors or hepatic tumors that are associated with other exposures. In rats, the substitution mutations found at A:T base pairs in the ras and p53 genes are consistent with the promutagenic properties of the DNA adduct 1,N(6)-ethenoadenine formed from VC metabolites. Risk assessments derived from animal studies seem to overestimate the actual risk of cancer when comparing estimated and reported cases of ASL.
Product Description
Supelco
Vinyl chloride solution, 200 μg/mL in methanol, analytical standard
Supelco
Vinyl chloride solution, certified reference material, 2000 μg/mL in methanol