Merck
  • Influence of nanoparticle concentration on thermo-physical properties of CuO-propylene glycol nanofluids.
Influence of nanoparticle concentration on thermo-physical properties of CuO-propylene glycol nanofluids.
Journal of nanoscience and nanotechnology (2014-04-18)
Kuppusamy Swaminathan Suganthi, Anju K Radhakrishnan, Natarajan Anusha, Kalpoondi Sekar Rajan
ABSTRACT
Experiments were performed on the preparation and characterization of CuO-propylene glycol nanofluids. The influence of nanoparticle concentration and temperature on nanofluid viscosity reveals existence of a range of nanoparticle concentration and temperature in which the viscosity of nanofluid is lower than that of propylene glycol, possibly due to interactions between nanoparticles and propylene glycol. A temperature-independent, thermal conductivity enhancement of 38% was obtained for nanoparticle concentration of 1.5 vol% over a temperature range of 10-60 degrees C. We believe that particle clustering contributes to the thermal conductivity enhancement in CuO-propylene glycol nanofluids.
Product Description
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
(R)-(−)-1,2-Propanediol, 96%
Copper, sphere, 100pcs, diameter 3.0mm, precision sphere grade 200
Copper, tube, 1000mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 0.95mm, inside diameter 0.29mm, wall thickness 0.33mm, as drawn, 99.9%
Copper, microfoil, disks, 25mm, thinness 0.25μm, specific density 219.9μg/cm2, permanent mylar 3.5μm support, 99.99+%
Copper, rod, 100mm, diameter 6.35mm, as drawn, 99.999%
Copper, rod, 50mm, diameter 12.7mm, as drawn, 99.99+%
Copper, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Copper, tube, 1000mm, outside diameter 1.0mm, inside diameter 0.24mm, wall thickness 0.38mm, as drawn, 99.9%
Copper, rod, 50mm, diameter 6.35mm, as drawn, 99.999%