Merck
  • Nuclear magnetic resonance relaxation characterisation of water status of developing grains of maize (Zea mays L.) grown at different nitrogen levels.
Nuclear magnetic resonance relaxation characterisation of water status of developing grains of maize (Zea mays L.) grown at different nitrogen levels.
Journal of bioscience and bioengineering (2013-11-19)
Prameela Krishnan, Usha Kiran Chopra, Ajay Pal Singh Verma, Devendra Kumar Joshi, Ishwar Chand
ABSTRACT
Changes in water status of developing grains of maize (Zea mays L.) grown under different nitrogen levels were characterized by nuclear magnetic resonance (NMR) spectroscopy. There were distinct changes in water status of grains due to the application of different levels of nitrogen (0, 120 and 180 kg N ha(-1)). A comparison of the grain developmental characteristics, composition and physical properties indicated that, not only the developmental characteristics like grain weight, grain number/ear, and rate of grain filling increased, but also bound water characterized by the T2 component of NMR relaxation increased with nitrogen application (50-70%) and developmental stages leading to maturation (10-60%). The consistency in the patterns of responses to free water and intermediate water to increasing levels of nitrogen application and grain maturity suggested that nitrogen application resulted in more proportion of water to both bound- and intermediate states and less in free state. These changes are further corroborated by the concomitant increases in protein and starch contents in grains from higher nitrogen treatments as macromolecules like protein and starch retain more amount of water in the bound state. The results of the changes in T2 showed that water status during grain development was not only affected by developmental processes but also by nitrogen supply to plants. This study strongly indicated a clear nutrient and developmental stage dependence of grain tissue water status in maize.
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Sigma-Aldrich
Starch, from potato, acc. to Zulkowsky (starch, treated with glycerol at 190°C)