Merck
  • Low cost single-step purification of endoglucanase from Aspergillus fumigatus ABK-9.
Low cost single-step purification of endoglucanase from Aspergillus fumigatus ABK-9.
Indian journal of experimental biology (2014-01-15)
Arpan Das, Tanmay Paul, Suman K Halder, Arijit Jana, Kuntal Ghosh, Chiranjit Maity, Pradeep K Das Mohapatra, Bikas R Pati, Keshab C Mondal
ABSTRACT
Low cost agro-waste was used as adsorption support for single-step purification of endoglucanase from the culture filtrate of A. fumigatus ABK-9. Among various agro-waste substrates, 1% NaOH pretreated rice bran was proved to be the best for adsorbing about 74.8 and 71.1% of endoglucanase at 4 degrees C and 10 degrees C respectively. Langmuir type adsorption isotherm at 4 degrees C showed maximum adsorption of enzyme at pH 5.0, which was in the range of optimum pH of the enzyme. The rice bran column bound enzyme was maximally eluted by a mixture of acetate buffer (0.05 M, pH 5.5) and ethanol (40%, v/v) at a ratio of 3:2 and a flow rate of 1 mL/min. A 5.52-fold purification of the enzyme was achieved from culture supernatant. The specific activity and recovery yield after purification were 294.0 U/mg and 40.15%, respectively, which were comparable with other contemporary protocols. The homogeneity of the enzyme was tested through sodium dodecyl sulphate polyacrylamide gel electrophoresis and a single band of 56.3 kDa was observed. Zymogram analysis finally confirmed the occurrence of endoglucanase in the single band.
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Cellulase from Aspergillus sp., aqueous solution
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid