Merck
  • Propofol compared with isoflurane inhibits mitochondrial metabolism in immature swine cerebral cortex.
Propofol compared with isoflurane inhibits mitochondrial metabolism in immature swine cerebral cortex.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2014-01-09)
Masaki Kajimoto, Douglas B Atkinson, Dolena R Ledee, Ernst-Bernhard Kayser, Phil G Morgan, Margaret M Sedensky, Nancy G Isern, Christine Des Rosiers, Michael A Portman
ABSTRACT
Anesthetics used in infants and children are implicated in the development of neurocognitive disorders. Although propofol induces neuroapoptosis in developing brain, the underlying mechanisms require elucidation and may have an energetic basis. We studied substrate utilization in immature swine anesthetized with either propofol or isoflurane for 4 hours. Piglets were infused with 13-Carbon-labeled glucose and leucine in the common carotid artery to assess citric acid cycle (CAC) metabolism in the parietal cortex. The anesthetics produced similar systemic hemodynamics and cerebral oxygen saturation by near-infrared spectroscopy. Compared with isoflurane, propofol depleted ATP and glycogen stores. Propofol decreased pools of the CAC intermediates, citrate, and α-ketoglutarate, while markedly increasing succinate along with decreasing mitochondrial complex II activity. Propofol also inhibited acetyl-CoA entry into the CAC through pyruvate dehydrogenase, while promoting glycolytic flux with marked lactate accumulation. Although oxygen supply appeared similar between the anesthetic groups, propofol yielded a metabolic phenotype that resembled a hypoxic state. Propofol impairs substrate flux through the CAC in the immature cerebral cortex. These impairments occurred without systemic metabolic perturbations that typically accompany propofol infusion syndrome. These metabolic abnormalities may have a role in the neurotoxity observed with propofol in the vulnerable immature brain.
Product Description
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-phospho-PDHE1-A type I (Ser293) Antibody, from rabbit, purified by affinity chromatography