Merck
  • Esterase activity in excised and reconstructed human skin--biotransformation of prednicarbate and the model dye fluorescein diacetate.
Esterase activity in excised and reconstructed human skin--biotransformation of prednicarbate and the model dye fluorescein diacetate.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2012-12-04)
Franzisca Marie Bätz, Wiebke Klipper, Hans Christian Korting, Frank Henkler, Robert Landsiedel, Andreas Luch, Uwe von Fritschen, Günther Weindl, Monika Schäfer-Korting
ABSTRACT
Reconstructed human epidermis (RHE) is used in non-animal testing for hazard analysis and reconstructed human skin (RHS) gains growing interest in preclinical drug development. RHE and RHS have been characterised regarding their barrier function, but knowledge about biotransformation capacity in these constructs and in human skin remains rather poor. However, metabolising enzymes can be highly relevant for the efficacy of topical dermatics as well as genotoxicity and sensitisation. We have compared the esteratic cleavage of the prednisolone diester prednicarbate and the enzyme kinetic parameters (Vmax and S0.5) of the model substrate fluorescein diacetate (FDA) in commercially available RHS and RHE with excised human skin and monolayer cultures of normal and immortalised human keratinocytes and of fibroblasts. Formation of the main metabolite prednisolone and of fluorescein ranked as: RHS~RHE>excised human skin and keratinocytes>fibroblasts, respectively. Because of the aromatic probe, however, Vmax of FDA cleavage did not show a linear relationship with prednicarbate metabolism. In conclusion, RHE and RHS may be useful to quantitatively address esterase activity of human skin in drug development and hazard analysis, although an increased activity compared to native human skin has to be taken into account.
Product Description
Prednicarbate for system suitability A, European Pharmacopoeia (EP) Reference Standard
Prednicarbate, European Pharmacopoeia (EP) Reference Standard