- Long-term efficacy and safety of firocoxib in the treatment of dogs with osteoarthritis.
Long-term efficacy and safety of firocoxib in the treatment of dogs with osteoarthritis.
The Veterinary record (2011-06-07)
A Autefage, F M Palissier, E Asimus, C Pepin-Richard
PMID21642297
ABSTRACT
Thirty-nine client-owned dogs with osteoarthritis were treated with 5 mg/kg firocoxib administered orally, once a day for 52 weeks. Veterinary examinations were performed on approximately days 0, 15, 90, 180, 270 and 360. Twenty-five dogs completed the study. The withdrawal rate associated with gastrointestinal side effects was low (5.1 per cent of dogs). Based on the owners' assessment, 82 per cent of the dogs had improved at day 15, 84 per cent of the 32 remaining dogs had improved at day 90, and 96 per cent of the 25 dogs that completed the trial had improved at day 360. During this trial, 12 (48 per cent) of the 25 remaining dogs showed an improvement in their lameness from day 90 to day 360 (P<0.05).
MATERIALS