- [Study on the chemical constituents of Uvaria microcarpa].
[Study on the chemical constituents of Uvaria microcarpa].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2009-10-31)
Ru-Mei Lu, Xing Su, Yuan-Yuan Zhou, Jian-Hua Wei
PMID19873731
ABSTRACT
To study the chemical constituents of Uvaria microcarpa. The constituents were repeatedly separated and purified with silica gel column and Sephadex LH-20 column, and identified by physico-chemical properties and spectral methods. Nine compounds were separated and identified as beta-sitosterol palmitate (I), euphorginol (II), beta-sitosterol (III), benzoic acid(IV), stigmasterol-3-O-beta-D-glucopyranoside (V), taraxerol (VI), emodin (VII), aristololactam A II (VI), beta-daucosterol (IX) . Compounds I, II, V, V are isolated from this plant for the first time.
MATERIALS