Merck
  • Transcriptional and post-transcriptional regulation of iNOS expression in human chondrocytes.
Transcriptional and post-transcriptional regulation of iNOS expression in human chondrocytes.
Biochemical pharmacology (2009-10-27)
Nadine Schmidt, Andrea Pautz, Julia Art, Peter Rauschkolb, Matthias Jung, Gerhard Erkel, Mary B Goldring, Hartmut Kleinert
ABSTRACT
Chondrocytes are important for the development and maintenance of articular cartilage. However, both in osteoarthritis (OA) and rheumatoid arthritis (RA) chondrocytes are involved in the process of cartilage degradation and synthesize important immunomodulatory mediators, including nitric oxide (NO) generated by the inducible NO synthase (iNOS). To uncover the role of iNOS in the pathomechanisms of OA and RA, we analyzed the regulation of iNOS expression using immortalized human chondrocytes as a reproducible model. In C-28/I2 chondrocytes, iNOS expression was associated with the expression of the chondrocyte phenotype. Peak induction by a cytokine cocktail occurred between 6 and 8h and declined by 24h. Inhibition of p38MAPK, NF-kappaB and the JAK2-STAT-1alpha pathways resulted in a reduction of iNOS expression. In contrast to other cell types, the cytokine-mediated induction of the human iNOS promoter paralleled the induction rate of the iNOS mRNA expression in C-28/I2 chondrocytes. However, in addition post-transcriptional regulation of iNOS expression by the RNA binding protein KSRP seems to operate in these cells. As seen in other chondrocyte models, glucocorticoids were not able to inhibit cytokine-induced iNOS expression in C-28/I2 cells, due to the lack of the glucocorticoid receptor mRNA expression. In this model of glucocorticoid-resistance, the new fungal anti-inflammatory compound S-curvularin was able to inhibit cytokine-induced iNOS expression and iNOS-dependent NO-production. In summary, we demonstrate for the first time that differentiated human immortalized C-28/I2 chondrocytes are a representative cell culture model to investigate iNOS gene expression in human joint diseases.
Sigma-Aldrich
Curvularin, from Penicillium citrinum, ≥98% (HPLC)
Pricing and availability is not currently available.