Merck
  • Simultaneous determination of paracetamol, caffeine and propyphenazone in ternary mixtures by micellar electrokinetic capillary chromatography.
Simultaneous determination of paracetamol, caffeine and propyphenazone in ternary mixtures by micellar electrokinetic capillary chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-10-19)
Deniz Emre, Nuran Ozaltin
ABSTRACT
A new micellar electrokinetic capillary chromatographic method has been developed to analyze the pharmaceutical preparations containing ternary combination of paracetamol (PAR), caffeine (CAF) and propyphenazone (PRO). Best results were obtained by using 20mM pH 9.0 borate buffer containing 30mM sodiumdodecylsulphate as the background electrolyte. Diflunisal (DIF) was used as internal standard (IS). The separation was performed through a fused silica capillary (50microm internal diameter, 44cm total length, 35.5cm effective length) at 25 degrees C with the application of 3s of hydrodynamic injection at 50mbar pressure and a potential of 29kV. Detection wavelength was 200nm. Under these conditions, the migration times were found to be 5.174min for PAR, 5.513min for CAF, 7.195min for DIF, and 9.366min for PRO. Linearity ranges for the method were determined as 2-200microgmL(-1) for PAR and CAF and 3-200microgmL(-1) for PRO. Limit of detections were found as 0.6microgmL(-1) for PAR and CAF and 0.8microgmL(-1) for PRO. According to the validation study, the developed method was proved to be accurate, precise, sensitive, specific, rugged and robust. Three pharmaceutical preparations, which are produced by different drug companies in Turkey, were analyzed by the developed method. One of the same preparations was also analyzed by the derivative ratio spectro zero-crossing spectrophotometric method reported in literature. No significant differences were found statistically.
Product Description
Propyphenazone, European Pharmacopoeia (EP) Reference Standard
Supelco
Propyphenazone, Pharmaceutical Secondary Standard; Certified Reference Material