Merck
  • Microflora changes with norfloxacin and pivmecillinam in women with recurrent urinary tract infection.
Microflora changes with norfloxacin and pivmecillinam in women with recurrent urinary tract infection.
Antimicrobial agents and chemotherapy (2006-03-30)
Birgit Stattin Norinder, Ragnar Norrby, Ann-Chatrin Palmgren, Sofia Hollenberg, Ulla Eriksson, Carl Erik Nord
ABSTRACT
Similar changes in the periurethral and vaginal microflora were observed in 19 women with recurrent urinary tract infection following treatment with norfloxacin (NOR) or pivmecillinam (PIV). Escherichia coli strains were suppressed by both treatments. Staphylococcus spp. and enterococci colony counts increased following PIV treatment in the periurethral flora but remained stable with NOR.
Sigma-Aldrich
Pivmecillinam, ≥98% (HPLC)