Merck
  • Detection and identification of phenazone-type drugs and their microbial metabolites in ground and drinking water applying solid-phase extraction and gas chromatography with mass spectrometric detection.
Detection and identification of phenazone-type drugs and their microbial metabolites in ground and drinking water applying solid-phase extraction and gas chromatography with mass spectrometric detection.
Journal of chromatography. A (2004-10-29)
Sebastian Zühlke, Uwe Dünnbier, Thomas Heberer
ABSTRACT
A new analytical method applying in situ derivatization was developed to enable the extraction of polar drug metabolites from water samples by solid-phase extraction (SPE). An additional derivatization by silylation was used to enhance the sensitivity of analyte detection by gas chromatography-mass spectrometry (GC-MS). Thus, the two metabolites 1,5-di-methyl-1,2-dehydro-3-pyrazolone (DP) and 4-(2-methylethyl)-1,5-dimethyl-1,2-dehydro-3-pyrazolone (PDP), postulated for the degradation of phenazone and propyphenazone, were identified and detected up to the microg/L level in raw and drinking water samples from public water supply.
Product Description
Supelco
Propyphenazone, Pharmaceutical Secondary Standard; Certified Reference Material
Propyphenazone, European Pharmacopoeia (EP) Reference Standard