- One step conversion of toxic beta-asarone from Acorus calamus into 1-(2,4,5-trimethoxyphenyl)-1,2-dihydroxypropane and asaronaldehyde occurring in Piper clusii.
One step conversion of toxic beta-asarone from Acorus calamus into 1-(2,4,5-trimethoxyphenyl)-1,2-dihydroxypropane and asaronaldehyde occurring in Piper clusii.
Natural product letters (2002-02-13)
A K Sinha, B P Joshi, R Dogra
PMID11838983
ABSTRACT
1-(2,4,5-Trimethoxyphenyl)-1,2-dihydroxypropane (2), a natural phenylpropanoid occurring in Piper clusii, has been synthesized for the first time from toxic beta-asarone (1) of Acorus calamus with osmium tetroxide, while 1 with osmium tetroxide (catalytic amount) in presence of sodium metaperiodate furnished the asaronaldehyde (3) in high yield.