Merck
  • Supramolecular concepts and new techniques in mechanochemistry: cocrystals, cages, rotaxanes, open metal-organic frameworks.
Supramolecular concepts and new techniques in mechanochemistry: cocrystals, cages, rotaxanes, open metal-organic frameworks.
Chemical Society reviews (2012-03-01)
Tomislav Friščić
ABSTRACT
Mechanochemical reactions effected by milling or grinding are an attractive means to conduct chemical reactions dependent on molecular recognition and to systematically explore different modes of molecular self-assembly. The natural relationship between milling mechanochemistry and supramolecular chemistry arises primarily from the ability to avoid bulk solvent, which simultaneously avoids limitations of solution-based chemistry, such as solubility, solvent complexation, or solvolysis, and makes the resulting process highly environmentally friendly. This tutorial review highlights the use of mechanochemistry for the synthesis of supramolecular targets in the solid state, such as molecular hydrogen- or halogen-bonded complexes, molecular and supramolecular cages, open frameworks and interlocked architectures. It is also demonstrated that the molecular self-assembly phenomena that are well-established in solution chemistry, such as reversible binding through covalent or non-covalent bonds, thermodynamic equilibration and structure templating, are also accessible in milling mechanochemistry through recently developed highly efficient methodologies such as liquid-assisted grinding (LAG) or ion- and liquid-assisted grinding (ILAG). Also highlighted are the new opportunities arising from the marriage of concepts of supramolecular and mechanochemical synthesis, including organocatalysis, deracemisation and discovery of new molecular recognition motifs.
Sigma-Aldrich
Bis(4-chlorophenyl) disulfide, 97%
Sigma-Aldrich
Bis(2-nitrophenyl) disulfide, 99%