Merck
  • Synthesis and evaluation of 2-substituted-6-phenyl-4,5-dihydropyridazin-3(2H)-ones as potent inodilators.
Synthesis and evaluation of 2-substituted-6-phenyl-4,5-dihydropyridazin-3(2H)-ones as potent inodilators.
Acta pharmaceutica (Zagreb, Croatia) (2008-12-24)
Dinesh Kumar, Rosalia Carron, Carmen De La Calle, Dharam Paul Jindal, Ranju Bansal
ABSTRACT
The present study describes the synthesis and pharmacological evaluation of 2-substituted-6-(4-acylaminophenyl)-4,5-dihydropyridazin-3(2H)-ones as potent inodilating agents. The synthesis of target compounds 2-4 and 7-11 was achieved by Friedel-Crafts acylation of appropriate anilide derivative with succinic anhydride or methylsuccinic anhydride and subsequent cyclization of intermediary keto acids with various hydrazine derivatives. The newly synthesized pyridazinone derivatives were evaluated for cardiotonic activity using isolated rat atria and for vasorelaxant activity using descending thoracic aortic rings of Wistar rats precontracted with phenylephrine (10-6 mol L-1). 6-(4-Methanesulfonamidophenyl)-2-phenyl-4,5-dihydropyridazin-3(2H)-one (7) exhibited significant inodilatory properties and showed vasorelaxant activity in a nanomolar range (IC50 = 0.08 +/- 0.01 mumol L-1).
Sigma-Aldrich
2-Hydrazino-2-imidazoline hydrobromide, 98%